Skip to Content
Merck

606421

Sigma-Aldrich

Acrolein-2-13C

≥99 atom % 13C, ≥90% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenal-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=13CHCHO
Molecular Weight:
57.06
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:


Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99 atom % 13C

Assay

≥90% (CP)

contains

hydroquinone as stabilizer

impurities

≤10% water

bp

53 °C (lit.)

mp

-87 °C (lit.)

density

0.850 g/mL at 25 °C

mass shift

M+1

storage temp.

−20°C

SMILES string

C=[13CH]C=O

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
M8250M293369892
MES low moisture content, ≥99% (titration)

M3671

MES

application(s)

clinical research
diagnostic assay manufacturing
life science and biopharma (
)
metabolomics (
)

application(s)

agriculture
clinical research
diagnostic assay manufacturing
life science and biopharma

application(s)

agriculture
diagnostic assay manufacturing
life science and biopharma
sample preparation

application(s)

agriculture
diagnostic assay manufacturing
general analytical
microbiology

form

powder

form

crystalline powder

form

crystalline powder

form

powder or crystals

density

1.23 g/cm3 at 20—25 °C, 1.27 g/cm3 at 20—25 °C

density

-

density

-

density

10.66 g/mL

storage condition

dry at room temperature

storage condition

dry at room temperature

storage condition

dry at room temperature

storage condition

-

pH

2.5-4 (25 °C, 97.6 g/L)

pH

-

pH

-

pH

-

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service