Skip to Content
Merck
All Photos(1)

Key Documents

A24001

Sigma-Aldrich

Acrolein diethyl acetal

96%

Synonym(s):

3,3-Diethoxy-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH(OCH2CH3)2
CAS Number:
Molecular Weight:
130.18
Beilstein:
1701567
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

125 °C (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)C=C

InChI

1S/C7H14O2/c1-4-7(8-5-2)9-6-3/h4,7H,1,5-6H2,2-3H3

InChI key

MCIPQLOKVXSHTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acrolein diethyl acetal is widely used to carry out chemoselective Heck arylation to synthesize either 3-arylpropanoate esters or cinnamaldehyde derivatives.

It can also be used as one of the precursors to synthesize natural products like (−)-(Z)-Deoxypukalide, (−)-Laulimalide, botryodiplodin, neolaulimalide and isolaulimalide.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient palladium-catalyzed synthesis of cinnamaldehydes from acrolein diethyl acetal and aryl iodides and bromides.
Battistuzzi G, et al.
Organic Letters, 5(5), 777-780 (2003)
Stefano Parisotto et al.
Organic & biomolecular chemistry, 15(4), 884-893 (2017-01-04)
As part of our ongoing work on the synthesis of a new class of plant hormones named Strigolactones (SLs) and their analogues, we became interested in tracing bioactive molecules with red emitting BODIPY fluorophores in order to unravel signaling and
Mohammad Saidur Rhaman et al.
Plant & cell physiology, 61(5), 967-977 (2020-03-08)
Myrosinase (β-thioglucoside glucohydrolase, enzyme nomenclature, EC 3.2.1.147, TGG) is a highly abundant protein in Arabidopsis guard cells, of which TGG1 and TGG2 function redundantly in abscisic acid (ABA)- and methyl jasmonate-induced stomatal closure. Reactive carbonyl species (RCS) are α,β-unsaturated aldehydes
Synthesis of (?)-and (−)-botryodiplodin using stereoselective radical cyclizations of acyclic esters and acetals.
Nouguier R et al.
Tetrahedron Asymmetry, 14(19), 3005-3018 (2003)
Total Synthesis of Microtubule-Stabilizing Agent (-)-Laulimalide1.
Ghosh AK et al.
The Journal of Organic Chemistry, 66(26), 8973-8982 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service