Skip to Content
Merck
All Photos(1)

Key Documents

595888

Sigma-Aldrich

3-(Trimethoxysilyl)propyl-N,N,N-trimethylammonium-15N chloride

99 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H24Cl15NO3Si
Molecular Weight:
258.82
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

isotopic purity

99 atom % 15N

Assay

97% (CP)

form

solid

mp

163 °C (lit.)

mass shift

M+1

SMILES string

[Cl-].CO[Si](CCC[N+](C)(C)C)(OC)OC

InChI

1S/C9H24NO3Si.ClH/c1-10(2,3)8-7-9-14(11-4,12-5)13-6;/h7-9H2,1-6H3;1H/q+1;/p-1

InChI key

FYZFRYWTMMVDLR-UHFFFAOYSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Frank et al.
Biological chemistry Hoppe-Seyler, 367(7), 573-578 (1986-07-01)
A new method for activation of glass fiber supports for immobilisation of proteins and peptides in gas-liquid phase sequencing is described. The new support offers several advantages over the presently used carrier polybrene: no precycling is required, initial yields are
Synthesis of a DNA-silica complex with rare two-dimensional square p4mm symmetry.
Chenyu Jin et al.
Angewandte Chemie (International ed. in English), 48(49), 9268-9272 (2009-11-10)
Huibin Qiu et al.
The journal of physical chemistry. B, 112(34), 10466-10474 (2008-08-02)
A series of achiral cationic and anionic amphiphiles employed as templates for mesoporous silica, all showed an excellent ability to form highly ordered helical mesoporous silica (HMS) crystals. It was supposed that the helical mesostructure may originate from the helical
Tatineni Balaji et al.
The Analyst, 130(8), 1162-1167 (2005-07-16)
A low cost, solid optical sensor for the rapid detection of low concentrations of Hg2+ in aqueous media was prepared by the monolayer functionalization of mesoporous silica with 5,10,15,20-tetraphenylporphinetetrasulfonic acid (TPPS), anchored by N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride (TMAC). The detection is based
Chiara Spagnoli et al.
Carbohydrate research, 340(5), 929-941 (2005-03-23)
Extended, relaxed, condensed, and interacting forms of the polysaccharide hyaluronan have been observed by atomic force microscopy (AFM). The types of images obtained depend on the properties of the surfaces used. We have investigated several different surface conditions for HA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service