Skip to Content
Merck
All Photos(1)

Key Documents

589764

Sigma-Aldrich

1-Tetradecan-d29-ol

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)12CD2OH
CAS Number:
Molecular Weight:
243.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

mp

38-40 °C (lit.)

mass shift

M+29

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2

InChI key

HLZKNKRTKFSKGZ-AUFKXYATSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service