Recommended Products
isotopic purity
97 atom % 15N
Assay
97% (CP)
form
solid
technique(s)
bio NMR: suitable
application(s)
peptide synthesis
functional group
Boc
Fmoc
mass shift
M+2
SMILES string
CC(C)(C)OC(=O)[15n]1cc(C[C@H]([15NH]C(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)c5ccccc15
InChI
1S/C31H30N2O6/c1-31(2,3)39-30(37)33-17-19(20-10-8-9-15-27(20)33)16-26(28(34)35)32-29(36)38-18-25-23-13-6-4-11-21(23)22-12-5-7-14-24(22)25/h4-15,17,25-26H,16,18H2,1-3H3,(H,32,36)(H,34,35)/t26-/m0/s1/i32+1,33+1
InChI key
ADOHASQZJSJZBT-QYIMINGJSA-N
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service