Skip to Content
Merck
All Photos(1)

Key Documents

579440

Sigma-Aldrich

Bis(2,2,2-trifluoroethyl) methylphosphonate

98%

Synonym(s):

Bis(2,2,2-trifluoroethyl) P-methylphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CH2O)2P(O)CH3
CAS Number:
Molecular Weight:
260.07
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.339 (lit.)

bp

183-184 °C (lit.)

mp

24-28 °C (lit.)

functional group

fluoro
phosphonate

SMILES string

CP(=O)(OCC(F)(F)F)OCC(F)(F)F

InChI

1S/C5H7F6O3P/c1-15(12,13-2-4(6,7)8)14-3-5(9,10)11/h2-3H2,1H3

InChI key

YIUFTMLPQFZEFD-UHFFFAOYSA-N

Application

Flame-retardant additive for lithium-ion electrolytes

Used as homogenous organic catalytic system and as a catalyst for phosphorylation of polyfluoroalkanols

Reactant for:
  • Synthesis of fluorescently labeled discodermolide for studying its binding to tubulin
  • Cross metathesis / phosphonate-based intramolecular olefination
  • Olefination for the synthesis of the anticancer microtubule-stabilizing macrolide, (-)-dictyostatin

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service