Skip to Content
Merck

576492

Sigma-Aldrich

Tributylphosphine tetrafluoroborate

97%

Synonym(s):

Tributylphosphonium tetrafluoroborate, [n-Bu3PH]BF4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3P · HBF4
CAS Number:
Molecular Weight:
290.13
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

50-53 °C (lit.)

functional group

phosphine

SMILES string

F[B-](F)(F)F.[H][P+](CCCC)(CCCC)CCCC

InChI

1S/C12H27P.BF4/c1-4-7-10-13(11-8-5-2)12-9-6-3;2-1(3,4)5/h4-12H2,1-3H3;/q;-1/p+1

InChI key

NCHAZLRRIGGCER-UHFFFAOYSA-O

General description

May contain approximately 5% of other butyl isomers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service