Skip to Content
Merck
All Photos(1)

Key Documents

558249

Sigma-Aldrich

Praseodymium(III) oxide

99.9% trace metals basis

Synonym(s):

Dipraseodymium trioxide, Praseodymia, Praseodymium oxide, Praseodymium sesquioxide, Praseodymium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr2O3
CAS Number:
Molecular Weight:
329.81
EC Number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

SMILES string

O=[Pr]O[Pr]=O

InChI

1S/3O.2Pr

InChI key

XYFIPBUCKXDCQM-UHFFFAOYSA-N

Application


  • Controlling the physics and chemistry of binary and ternary praseodymium and cerium oxide systems: This article discusses the progress in studying single crystalline praseodymium and cerium oxide materials and their characterization methods (G Niu, MH Zoellner, T Schroeder, A Schaefer, 2015).

  • A close look at the influence of praseodymium (III) oxide on the structural, physical, and gamma-ray protection capacity of a ternary B2O3 PbO CdO glass system: Explores the role of Praseodymium(III) oxide in enhancing the properties of heavy metal oxide glass systems, particularly in radiation shielding (RH Shoeir, M Afifi, AS Dhmees, MI Sayyed, 2024).

  • Praseodymium oxide nanostructures: novel solvent-less preparation, characterization and investigation of their optical and photocatalytic properties: Discusses a new method for synthesizing praseodymium oxide nanostructures, examining their optical and photocatalytic activities (S Zinatloo-Ajabshir, M Salavati-Niasari, 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service