Skip to Content
Merck
All Photos(3)

Key Documents

634611

Sigma-Aldrich

Neodymium(III) oxide

nanopowder, <100 nm particle size (BET), 99.9% trace metals basis

Synonym(s):

Dineodymium trioxide, Neodymia, Neodymium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd2O3
CAS Number:
Molecular Weight:
336.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: neodymium

particle size

<100 nm (BET)

density

7.24 g/mL at 20 °C (lit.)

SMILES string

O=[Nd]O[Nd]=O

InChI

1S/2Nd.3O

InChI key

HBPPDPSYLUIKHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Chen et al.
International journal of nanomedicine, 15, 387-400 (2020-02-06)
Rare-earth nanoparticles in the environment and human body pose a potential threat to human health. Although toxic effects of rare-earth nanoparticles have been extensively studied, the effects on the early development are not well understood. In this study, we attempted

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service