Skip to Content
Merck
All Photos(1)

Key Documents

553441

Sigma-Aldrich

Tris(tert-pentoxy)silanol

≥99.99%

Synonym(s):

TPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2C(CH3)2O)3SiOH
CAS Number:
Molecular Weight:
306.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99%

form

liquid

bp

96-99 °C/2-3 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)O[Si](O)(OC(C)(C)CC)OC(C)(C)CC

InChI

1S/C15H34O4Si/c1-10-13(4,5)17-20(16,18-14(6,7)11-2)19-15(8,9)12-3/h16H,10-12H2,1-9H3

InChI key

ORJFXWYTRPGGRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfredo Mirandola et al.
Medical physics, 42(9), 5287-5300 (2015-09-04)
To describe the dosimetric commissioning and quality assurance (QA) of the actively scanned proton and carbon ion beams at the Italian National Center for Oncological Hadrontherapy. The laterally integrated depth-dose-distributions (IDDs) were acquired with the PTW Peakfinder, a variable depth
Hendrik Thoen et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 115(1), 16-21 (2015-03-31)
Neoadjuvant chemoradiotherapy is used prior to surgery in curative treatment of esophageal cancer (EC). We evaluated the in-field control of this multimodal treatment by extraction of radiation dose parameters and determination of the spatial relation between tumor recurrence location(s) and
Hao Jiang et al.
Oncology letters, 10(5), 2731-2738 (2016-01-02)
The aim of the present study was to retrospectively analyze the clinical efficacy and side-effects of two-dimensional conventional radiotherapy (2D-CRT) and intensity-modulated radiotherapy (IMRT) in 53 NPC patients with cervical spine involvement, without distant metastases. In total, 53 patients were
Eduardo S Moura et al.
Medical physics, 42(4), 1566-1574 (2015-04-04)
This work presents the development of a phantom to verify the treatment planning system (TPS) algorithms used for high-dose-rate (HDR) brachytherapy. It is designed to measure the relative dose in a heterogeneous media. The experimental details used, simulation methods, and
V E Bellinzona et al.
Physica medica : PM : an international journal devoted to the applications of physics to medicine and biology : official journal of the Italian Association of Biomedical Physics (AIFB), 31(5), 484-492 (2015-06-03)
The accurate evaluation of the lateral dose profile is an important issue in the field of proton radiation therapy. The beam spread, due to Multiple Coulomb Scattering (MCS), is described by the Molière's theory. To take into account also the

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service