Skip to Content
Merck
All Photos(2)

Key Documents

105198

Sigma-Aldrich

(+)-Dehydroabietylamine

technical grade, 60%

Synonym(s):

1,4a-Dimethyl-7-isopropyl-1,2,3,4,4a,9,10,10a-octahydro-1-phenanthrenemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31N
CAS Number:
Molecular Weight:
285.47
Beilstein:
3084620
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

optical activity

[α]20/D +56.1°, c = 2.4 in pyridine

composition

high molecular weight primary amines, 90%

concentration

60%

refractive index

n20/D 1.546 (lit.)

SMILES string

[H][C@@]12CCc3cc(ccc3[C@@]1(C)CCC[C@@]2(C)CN)C(C)C

InChI

1S/C20H31N/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4/h6,8,12,14,18H,5,7,9-11,13,21H2,1-4H3/t18-,19-,20+/m0/s1

InChI key

JVVXZOOGOGPDRZ-SLFFLAALSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Ayaz Mustufa et al.
BMC cancer, 16(1), 883-883 (2016-11-16)
The purpose of present work is to synthesize novel (+)-Dehydroabietylamine derivatives (DAAD) using N-acetyl-α-amino acid conjugates and determine its cytotoxic effects on hepatocellular carcinoma cells. An analytical study was conducted to explore cytotoxic activity of DAAD on hepatocellular carcinoma cell
Tiina Laaksonen et al.
Molecules (Basel, Switzerland), 20(11), 20873-20886 (2015-11-28)
Chiral tertiary and quaternary amine solvating agents for NMR spectroscopy were synthesized from the wood resin derivative (+)-dehydroabietylamine (2). The resolution of enantiomers of model compounds [Mosher's acid (3) and its n-Bu₄N salt (4)] (guests) by (+)-dehydroabietyl-N,N-dimethylmethanamine (5) and its

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service