Skip to Content
Merck
All Photos(1)

Key Documents

55220

Sigma-Aldrich

1-Butyl-3-methylimidazolium dicyanamide

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5
CAS Number:
Molecular Weight:
205.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

impurities

≤0.2% water

functional group

amine

SMILES string

N#C[N-]C#N.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.C2N3/c1-3-4-5-10-7-6-9(2)8-10;3-1-5-2-4/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

ICIVTHOGIQHZRY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Alekseeva et al.
Materials (Basel, Switzerland), 12(16) (2019-08-16)
Composites of montmorillonite K10 (MMT K10) and ionic liquid (IL) containing a 1-butyl-3-methyl-imidazolium cation ([BMIm]+) and various anions, such as bis (trifluoromethylsulfonyl) imide ([NTf2]-), trifluoromethanesulfonate ([OTf]-), and dicyanamide ([DCA]-) have been obtained in this work. A number of methods, such
Aafiya Tarannum et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 212, 343-348 (2019-01-23)
In protein-ionic liquids (ILs) interactions, anions play an important role. In this work, imidazolium-based ILs (IILs) with varying anions namely dicyanamide (DCA), hydrogen sulfate (HS), dimethyl phosphate (DP), acetate (A), sulfate (S) and dihydrogen phosphate (DHP) have been chosen with
Paula Navalpotro et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(10), 1800576-1800576 (2018-10-26)
Aqueous biphasic systems (ABS) formed by water, ionic liquids (ILs), and salts, in which the two phases are water rich, are demonstrated here to act as potential membrane-free batteries. This concept is feasible due to the selective enrichment of redox
Karolina Matuszek et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-28)
Low solubility of terephthalic acid in common solvents makes its industrial production very difficult and not environmentally benign. Ionic liquids are known for their extraordinary solvent properties, with capability to dissolve a wide variety of materials, from common solvents to
Lillian A Frink et al.
Journal of pharmaceutical and biomedical analysis, 94, 111-117 (2014-02-25)
A rapid, accurate, precise and versatile analytical method was developed for the detection and quantification of water in solid active pharmaceutical ingredients (APIs). The headspace gas chromatography (HSGC) method utilized an ionic liquid (IL) based open tubular capillary GC column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service