Merck
  • Water determination in active pharmaceutical ingredients using ionic liquid headspace gas chromatography and two different detection protocols.
Water determination in active pharmaceutical ingredients using ionic liquid headspace gas chromatography and two different detection protocols.
Journal of pharmaceutical and biomedical analysis (2014-02-25)
Lillian A Frink, Choyce A Weatherly, Daniel W Armstrong
ABSTRACT
A rapid, accurate, precise and versatile analytical method was developed for the detection and quantification of water in solid active pharmaceutical ingredients (APIs). The headspace gas chromatography (HSGC) method utilized an ionic liquid (IL) based open tubular capillary GC column to increase sensitivity and ruggedness of this method. ILs are also utilized as the headspace solvent because of their low vapor pressure, unique physiochemical properties and high thermal stability. This method is not affected by side reactions and solubility problems which are common with Karl Fischer Titration (KFT) methods. Nor is it as limited as weight loss on drying approaches. The ability to use either/both modern thermal conductivity or barrier ion discharge GC detection provides flexibility, different dynamic ranges and sensitivity. The developed method also was shown to be broadly applicable.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Warfarin, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
1-Butyl-3-methylimidazolium dicyanamide, ≥97.0% (HPLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
USP
Warfarin, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland