Skip to Content
Merck
All Photos(1)

Key Documents

549223

Sigma-Aldrich

3,4-Difluorothiophenol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3SH
CAS Number:
Molecular Weight:
146.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.528 (lit.)

bp

169-170 °C (lit.)

density

1.323 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc(S)cc1F

InChI

1S/C6H4F2S/c7-5-2-1-4(9)3-6(5)8/h1-3,9H

InChI key

BGVRHDQMTMPAEZ-UHFFFAOYSA-N

General description

3,4-Difluorothiophenol is a halo-substituted thiol derivative.

Application

3,4-Difluorothiophenol may be used:
  • To prepare a novel 3,4-difluorophenylthiomethyl group containing phthalazine derivative, which shows high cytotoxic activity.
  • To prepare a fluorothiophenyl modified poly(vinyl chloride) membrane with improved gas permeability efficiency.
  • As a ligand to synthesize noble metal quantum clusters (QCs).
  • To prepare ethyl 5-(aminosulfonyl)-2-[(3,4-difluorophenyl)thio]nicotinate, an intermediate for preparing potent thioether benzenesulfonamide inhibitors of carbonic anhydrases II and IV.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noble metal clusters: applications in energy, environment, and biology.
Mathew A and Pradeep T.
Particle & Particle Systems Characterization, 31(10), 1017-1053 (2013)
Synthesis and in vitro cytotoxicity of novel 1, 4-disubstituted phthalazines.
Zhai X, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 19(1), 29-32 (2008)
Thioether benzenesulfonamide inhibitors of carbonic anhydrases II and IV: structure-based drug design, synthesis, and biological evaluation.
Vernier W, et al.
Bioorganic & Medicinal Chemistry, 18(9), 3307-3319 (2010)
Gas transport in fluorothiophenyl modified PVC membranes.
Bierbrauer K, et al.
Journal of Membrane Science, 362(1), 164-171 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service