Skip to Content
Merck
All Photos(1)

Key Documents

545430

Sigma-Aldrich

3-Octyn-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C≡CCH2CH2OH
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4569 (lit.)

density

0.880 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCC#CCCO

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h9H,2-4,7-8H2,1H3

InChI key

LRZGRGVRZSDRTK-UHFFFAOYSA-N

General description

3-Octyn-1-ol is a homopropargylic alcohol that can be prepared from 3-butyn-1-ol and 1-bromobutane.

Application

3-Octyn-1-ol may be used in the preparation of:
  • (3Z)-octen-1-ol
  • 7-octyn-1-ol
  • 3-cis-octenoic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd/CaCO3 in liquid poly (ethylene glycol)(PEG): an easy and efficient recycle system for partial reduction of alkynes to cis-olefins under a hydrogen atmosphere.
Chandrasekhar S, et al.
Tetrahedron Letters, 45(11), 2421-2423 (2004)
Peter Witzgall et al.
Journal of chemical ecology, 31(12), 2923-2932 (2005-12-21)
Analysis of extracts of sex pheromone glands of grapevine moth females Lobesia botrana showed three previously unidentified compounds, (E)-7-dodecenyl acetate and the (E,E)- and (Z,E)-isomers of 7,9,11-dodecatrienyl acetate. This is the first account of a triply unsaturated pheromone component in
Selective cleavage of dimethylhydrazones to the carbonyl compounds using silica gel and its application in the synthesis of (Z)-9-tetradecenyl acetate.
Mitra RB and Reddy GB.
Synthesis, 1989(09), 694-698 (1989)
Dongyan Zhang et al.
The Journal of biological chemistry, 277(11), 9127-9132 (2002-01-10)
The degradation of unsaturated fatty acids by beta-oxidation involves Delta(3),Delta(2)-enoyl-CoA isomerases (enoyl-CoA isomerases) that catalyze 3-cis --> 2-trans and 3-trans --> 2-trans isomerizations of enoyl-CoAs and the 2,5 --> 3,5 isomerization of dienoyl-CoAs. An analysis of rat liver enoyl-CoA isomerases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service