Skip to Content
Merck
All Photos(1)

Key Documents

544027

Sigma-Aldrich

5,7-Hexadecadiynoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3C(CH2)7C≡C2≡C(CH2)3CO2H
CAS Number:
Molecular Weight:
248.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

44-48 °C (lit.)

SMILES string

CCCCCCCCC#CC#CCCCC(O)=O

InChI

1S/C16H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-8,13-15H2,1H3,(H,17,18)

InChI key

RPTNONNBJKKVEV-UHFFFAOYSA-N

General description

5,7-Hexadecadiynoic acid is a diacetylenic acid. It participates in the synthesis of polymerizable asymmetric diacetylenic monomer compounds (ADMs). The FT-Raman and FT-IR spectra of 5,7-hexadecadiynoic acid have been recorded and assigned.

Application

5,7-Hexadecadiynoic acid (HDDA) may be used in the preparation of poly(HDDA)/zinc oxide(ZnO) nanocomposites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Controlling the reversible thermochromism of polydiacetylene/zinc oxide nanocomposites by varying alkyl chain length.
Chanakul A, et al.
Journal of Colloid and Interface Science, 389(1), 106-114 (2013)
Towards a universal method for the stable and clean functionalization of inert perfluoropolymer nanoparticles: Exploiting photopolymerizable amphiphilic diacetylenes.
Morasso C, et al.
Advances in Functional Materials, 20(22), 3932-3940 (2010)
Dual colorimetric response of polydiacetylene/Zinc oxide nanocomposites to low and high pH.
Chanakul A, et al.
Journal of Colloid and Interface Science, 418, 43-51 (2014)
Colorimetric sensing of various organic acids by using polydiacetylene/zinc oxide nanocomposites: Effects of polydiacetylene and acid structures.
Chanakul A, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 489, 9-18 (2016)
Vibrational and theoretical study of diacetylenic acids.
Roman M and Baranska M.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 137, 652-660 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service