Skip to Content
Merck
All Photos(1)

Key Documents

793361

Sigma-Aldrich

Zinc oxide nanoparticle ink

Synonym(s):

Avantama N-10, Nanograde N-10, Zinc oxide suspension, ZnO dispersion, ZnO ink, ZnO nanoparticle ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion

Quality Level

concentration

2.5 wt. % (crystalline ZnO in 2-propanol)

color

translucent yellowish

particle size

10-15 nm

density

0.800 g/mL at 25 °C

General description

Zinc oxide nanoparticle ink are nanoparticle-based printing inks allowing processing temperatures of 80°C. Zinc oxide nanoparticle ink is an electron selective interface layer ink containing zinc oxide (ZnO) nanoparticles in isopropanol. ZnO nanoparticle ink functions well as electron extraction layer (EEL) materials in solar cells.

Application

This ZnO nanoparticle ink is for slot-dye, spin-coating, and doctor blading for the use as electron transport layer in printed electronics. This ZnO nanoparticle ink is universally applicable in normal and inverted architecture.
Annealing temperature <100°C.

Other Notes

Prior to application: Ultrasonicate and (optionally) filter through 0.45 μm PTFE filter
Post-treatment: Annealing of deposited ZnO films at 80°C - 120°C

Legal Information

Product of Avantama Ltd.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A universal method to form the equivalent ohmic contact for efficient solution-processed organic tandem solar cells
Journal of Material Chemistry A, 2, 14896?14902-14896?14902 (2014)
Chun-Chao Chen et al.
Advanced materials (Deerfield Beach, Fla.), 26(32), 5670-5677 (2014-07-22)
Tandem solar cells have the potential to improve photon conversion efficiencies (PCEs) beyond the limits of single-junction devices. In this study, a triple-junction tandem design is demonstrated by employing three distinct organic donor materials having bandgap energies ranging from 1.4
Flexible organic tandem solar modules with 6% efficiency: combining roll-to-roll compatible processing with high geometric fill factors
Energy & Environmental Science, 7, 3284?3290-3284?3290 (2014)
Interface Design to Improve the Performance and Stability of Solution-Processed Small-Molecule Conventional Solar Cells
Advanced Energy Materials (2014)

Articles

Find advantages of inorganic interface layer inks for organic electronic & other applications.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service