543292
IR-783
Dye content 90 %
Synonym(s):
2-[2-[2-Chloro-3-[2-[1,3-dihydro-3,3-dimethyl-1-(4-sulfobutyl)-2H-indol-2-ylidene]-ethylidene]-1-cyclohexen-1-yl]-ethenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium hydroxide, inner salt sodium salt
About This Item
Recommended Products
form
solid
composition
Dye content, 90%
mp
209-212 °C (lit.)
λmax
782 nm
SMILES string
[Na+].CC1(C)\C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCS([O-])(=O)=O)c4ccccc4C3(C)C)=C2Cl)N(CCCCS([O-])(=O)=O)c5ccccc15
InChI
1S/C38H47ClN2O6S2.Na/c1-37(2)30-16-5-7-18-32(30)40(24-9-11-26-48(42,43)44)34(37)22-20-28-14-13-15-29(36(28)39)21-23-35-38(3,4)31-17-6-8-19-33(31)41(35)25-10-12-27-49(45,46)47;/h5-8,16-23H,9-15,24-27H2,1-4H3,(H-,42,43,44,45,46,47);/q;+1/p-1
InChI key
QQIQAVJARACLHE-UHFFFAOYSA-M
General description
Application
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Graphene is the building block for carbon nanomaterials with different dimensionalities.
Graphene is the building block for carbon nanomaterials with different dimensionalities.
Graphene is the building block for carbon nanomaterials with different dimensionalities.
Graphene is the building block for carbon nanomaterials with different dimensionalities.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service