Skip to Content
Merck
All Photos(2)

Key Documents

425311

Sigma-Aldrich

IR-780 iodide

Dye content ≥95 %

Synonym(s):

2-[2-[2-Chloro-3-[(1,3-dihydro-3,3-dimethyl-1-propyl-2H-indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-3,3-dimethyl-1-propylindolium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44ClIN2
CAS Number:
Molecular Weight:
667.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Dye content, ≥95%

mp

232-234 °C (lit.)

λmax

780 nm

SMILES string

[I-].CCCN1c2ccccc2C(C)(C)/C1=C\C=C3/CCCC(\C=C\C4=[N+](CCC)c5ccccc5C4(C)C)=C3Cl

InChI

1S/C36H44ClN2.HI/c1-7-24-38-30-18-11-9-16-28(30)35(3,4)32(38)22-20-26-14-13-15-27(34(26)37)21-23-33-36(5,6)29-17-10-12-19-31(29)39(33)25-8-2;/h9-12,16-23H,7-8,13-15,24-25H2,1-6H3;1H/q+1;/p-1

InChI key

IRPKBYJYVJOQHQ-UHFFFAOYSA-M

General description

IR-780 iodide is a near-infrared lipophilic cation heptamethine fluorescent dye. It has a good fluorescent intensity and can be used as a photothermal therapy agent. It shows an optical absorption spectrum of 780 nm that makes it useful as a sonodynamic therapy agent for medical treatment.

Application

IR-780 based dyes can be potentially used in a biomedical application and drug delivery for tumor imaging and targeted therapy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IR-780 dye for near-infrared fluorescence imaging in prostate cancer
Yi X, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 21(28), 511-511 (2015)
IR-780 dye loaded tumor targeting theranostic nanoparticles for NIR imaging and photothermal therapy
Yue C, et al.
Biomaterials, 34(28), 6853-6861 (2013)
IR-780 dye as a sonosensitizer for sonodynamic therapy of breast tumor
Li Y, et al.
Scientific Reports, 6(28), 25968-25968 (2016)
Sentinel lymph node mapping by a near-infrared fluorescent heptamethine dye
Zhang C, et al.
Biomaterials, 31(7), 1911-1917 (2010)
Preferential accumulation of the near infrared heptamethine dye IR-780 in the mitochondria of drug-resistant lung cancer cells
Wang Y, et al.
Biomaterials, 35(13), 4116-4124 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service