Skip to Content
Merck
All Photos(1)

Key Documents

532711

Sigma-Aldrich

2-Fluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2Cl
CAS Number:
Molecular Weight:
194.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.537 (lit.)

bp

246-247 °C (lit.)

mp

27-30 °C (lit.)

density

1.47 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1S(Cl)(=O)=O

InChI

1S/C6H4ClFO2S/c7-11(9,10)6-4-2-1-3-5(6)8/h1-4H

InChI key

ZSZKAQCISWFDCQ-UHFFFAOYSA-N

General description

2-Fluorobenzenesulfonyl chloride, also known as o-fluorobenzenesulfonyl chloride, is a flurinated arylsulfonyl chloride. It can be prepared from o-benzenedisulfonyl fluoride.

Application

2-Fluorobenzenesulfonyl chloride may be used in the preparation of the following furan derivatives:
  • 2-(2-fluorophenyl)benzofuran
  • 2-butyl-5-(2-fluorophenyl)furan
  • 2-(2-fluorophenyl)-3,6-dimethyl-4,5,6,7-tetrahydrobenzofuran

It may also be used to prepare:
  • 2-fluorobenzenesulfonamide
  • methyl 2-{[(2-fluorophenyl)sulfonyl]amino}-5,6,7,8-tetrahydro-1-naphthalenecarboxylate
  • potassium fluorobenzene-2-sulfonate
  • 1-(2-bromobenzyl)-2-(2-fluorophenyl)pyrrole

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and optimization of anthranilic acid sulfonamides as inhibitors of methionine aminopeptidase-2: a structural basis for the reduction of albumin binding.
Sheppard GS, et al.
Journal of Medicinal Chemistry, 49(13), 3832-3849 (2006)
Regiocontroled Palladium?Catalysed Direct Arylation at Carbon C2 of Benzofurans using Benzenesulfonyl Chlorides as the Coupling Partners.
Loukotova L, et al.
ChemCatChem, 6(5), 1303-1309 (2014)
Water-Soluble Phosphines. 6.1 Tailor-Made Syntheses of Chiral Secondary and Tertiary Phosphines with Sulfonated Aromatic Substituents: Structural and Quantum Chemical Studies.
Bitterer F, et al.
Inorganic Chemistry, 35(14), 4103-4113 (1996)
Benzenesulfonyl Chlorides: Alternative Coupling Partners for Regiocontrolled Palladium-Catalyzed Direct Desulfitative 5-Arylation of Furans.
Beladhria A, et al.
Synthesis, 46(18), 2515-2523 (2014)
Palladium-Catalyzed Iterative C- H Bond Arylations: Synthesis of Medium-Size Heterocycles with a Bridgehead Nitrogen Atom.
Hagui W, et al.
ChemCatChem, 7(21), 3544-3554 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service