Skip to Content
Merck
All Photos(2)

Key Documents

523755

Sigma-Aldrich

2,3,5,6-Tetramethyl-p-phenylenediamine

electronic grade, 99% trace metals basis

Synonym(s):

3,6-Diaminodurene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4C6(NH2)2
CAS Number:
Molecular Weight:
164.25
Beilstein:
2208743
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Quality Level

Assay

99% trace metals basis

mp

150-155 °C (lit.)

SMILES string

Cc1c(C)c(N)c(C)c(C)c1N

InChI

1S/C10H16N2/c1-5-6(2)10(12)8(4)7(3)9(5)11/h11-12H2,1-4H3

InChI key

WCZNKVPCIFMXEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monomer used in the preparation of polyimide coatings for electronic applications-IC passivation, dielectric for multichip modules, and as a stress relief layer in sensor applications.

Other Notes

Contains low metals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew P Johnson et al.
The Journal of biological chemistry, 286(22), 19973-19981 (2011-04-09)
Variations in the light environment require higher plants to regulate the light harvesting process. Under high light a mechanism known as non-photochemical quenching (NPQ) is triggered to dissipate excess absorbed light energy within the photosystem II (PSII) antenna as heat
B K Kolli et al.
Indian journal of biochemistry & biophysics, 33(6), 465-470 (1996-12-01)
The kinetic parameters of different sites of electron donation to photosystem I (PS I) were evaluated in Spirulina platensis thylakoids. Reduced 2,6-dichlorophenolindophenol (DCIPH2) exhibited two sites of electron donation, with apparent K(m) values of 8 and 40 microM each. The
C Schwarze et al.
European journal of biochemistry, 267(2), 422-433 (2000-01-13)
Flash-induced redox changes of b-type and c-type cytochromes have been studied in chromatophores from the aerobic photosynthetic bacterium Roseobacter denitrificans under redox-controlled conditions. The flash-oxidized primary donor P+ of the reaction center (RC) is rapidly re-reduced by heme H1 (Em,7
K Sonoike et al.
FEBS letters, 362(2), 235-238 (1995-04-03)
The activity of photosystem (PS) I in cucumber leaves was selectively inhibited by weak illumination at chilling temperatures with almost no loss of P-700 content and PSII activity. The sites of inactivation in the reducing side of PSI were determined
Critical evaluation of the proton-translocating property of cytochrome oxidase in rat liver mitochondria.
M Wikström et al.
FEBS letters, 144(2), 183-189 (1982-08-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service