Skip to Content
Merck
All Photos(2)

Key Documents

386448

Sigma-Aldrich

4,4′-(Hexafluoroisopropylidene)diphthalic anhydride

99%

Synonym(s):

2,2-Bis(3,4-dicarboxyphenyl)-1,1,1,3,3,3-hexafluoropropane, 4,4′-(Hexafluoroisopropylidene)bis(phthalic anhydride), 4,4′-(Hexafluoroisopropylidene)diphthalic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H6F6O6
CAS Number:
Molecular Weight:
444.24
Beilstein:
7057916
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

244-247 °C (lit.)

SMILES string

FC(F)(F)C(c1ccc2C(=O)OC(=O)c2c1)(c3ccc4C(=O)OC(=O)c4c3)C(F)(F)F

InChI

1S/C19H6F6O6/c20-18(21,22)17(19(23,24)25,7-1-3-9-11(5-7)15(28)30-13(9)26)8-2-4-10-12(6-8)16(29)31-14(10)27/h1-6H

InChI key

QHHKLPCQTTWFSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn K Post et al.
Parkinsonism & related disorders, 14(7), 524-531 (2008-05-20)
Parkinson's disease (PD) has classically been characterized by features of motor dysfunction, but these may manifest only after the nigrostriatal system has incurred significant damage. However, recent evidence suggests that cardiac sympathetic denervation occurs early on in PD. This may
H Kawakami et al.
Journal of biomaterials science. Polymer edition, 7(12), 1029-1038 (1996-01-01)
Fluorinated polyimide derived from 2,2'-bis(3,4-dicarboxyphenyl) hexafluoropropane dianhydride (6FDA) and bis[4-(4-aminophenoxy) phenyl]sulfone (APPS) was synthesized to develop a novel membrane oxygenator combining excellent gas transfer and blood compatibility. The asymmetric gas exchange membranes of 6FDA-APPS made by a dry/wet process consisted
Yingyi Ma et al.
Chemistry, an Asian journal (2018-06-28)
A honeycomb-patterned porous film was conveniently prepared by means of the microemulsion method and then used as an effective coating for improving the dielectric properties and water resistance of polyimide (PI) film while maintaining its mechanical properties. The dielectric properties
Fidel E Rodríguez-González et al.
Membranes, 10(7) (2020-07-09)
A set of five new aromatic poly(imide)s (PIs) incorporating pendant acyclic alkyl moieties were synthesized. The difference among them was the length and bulkiness of the pendant group, which comprises of linear alkyl chains from three to six carbon atoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service