All Photos(1)
About This Item
Linear Formula:
(CH3)2C6H3CHO
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.538 (lit.)
bp
232 °C (lit.)
density
0.998 g/mL at 25 °C (lit.)
functional group
aldehyde
SMILES string
Cc1cc(C)cc(C=O)c1
InChI
1S/C9H10O/c1-7-3-8(2)5-9(4-7)6-10/h3-6H,1-2H3
InChI key
NBEFMISJJNGCIZ-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
210.2 °F - closed cup
Flash Point(C)
99.00 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Design and synthesis of novel chiral pyrrolidine-based diamines with C2-symmetry.
Nakajima M, et al.
Tetrahedron, 49(43), 9735-9750 (1993)
H Tanaka et al.
Journal of medicinal chemistry, 38(15), 2860-2865 (1995-07-21)
Several 6-benzyl analogs of 1-[(2-hydroxyethoxy)methyl]-6-(phenylthio)thymine (1; HEPT) were synthesized and evaluated for their anti-HIV-1 activity. LDA (lithium diisopropylamide) lithiation of 5-ethyluracil derivatives 7 and 8 and subsequent reaction with an aryl aldehyde gave 6-(arylhydroxymethyl)-5-ethyluracil derivatives 9-12. 6-(Arylhydroxymethyl)-5-isopropyluracil derivatives 15-18 were
A new substrate for the Biginelli cyclocondensation: direct preparation of 5-unsubstituted 3,4-dihydropyrimidin-2(1H)-ones from a beta-keto carboxylic acid
Bussolari et al.
The Journal of organic chemistry, 65(20), 6777-6779 (2000-10-29)
Kejing An et al.
Food chemistry, 301, 125282-125282 (2019-08-07)
The main contributors to the cooked off-flavor in heat-sterilized lychee juice (HLJ) were studied by means of molecular sensory science. The HLJ which elicited cooked cabbage/potato and onion/garlic off-flavor was compared with fresh lychee juice (FLJ) having desired sensory attributes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service