Skip to Content
Merck
All Photos(1)

Key Documents

511323

Sigma-Aldrich

Di-tert-butyl iminodiacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN[CH2CO2C(CH3)3]2
CAS Number:
Molecular Weight:
245.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

38-42 °C (lit.)

SMILES string

CC(C)(C)OC(=O)CNCC(=O)OC(C)(C)C

InChI

1S/C12H23NO4/c1-11(2,3)16-9(14)7-13-8-10(15)17-12(4,5)6/h13H,7-8H2,1-6H3

InChI key

SMXMBXPLRFTROI-UHFFFAOYSA-N

General description

Di-tert-butyl iminodiacetate is a secondary amine.

Application

Di-tert-butyl iminodiacetate may be used as a reagent in the synthesis of:
  • multi-carboxylic acid-containing carbocyanine dyes
  • monosubstituted difunctionalized polyhedral oligomeric silsesquioxanes (POSS) monomers
  • multigenerational fluorinated dendrimers
  • 2,6-dipyrazol-1-ylpyridine derivatives
It may also be used for the preparation of di-tert-butyl N-(4-allyloxy-4-oxobutanoyl)iminodiacetate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-Supported Synthesis of Bicyclic Peptides Containing Three Parallel Peptide Chains.
Karskela T, et al.
European Journal of Organic Chemistry, 2003(9), 1687-1691 (2003)
Fluorous mixture synthesis of asymmetric dendrimers.
Jiang ZX and Yu YB.
The Journal of Organic Chemistry, 75(6), 2044-2049 (2010)
A facile route to difunctionalized monosubstituted octasilsesquioxanes.
Liu H, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 17(18), 5019-5023 (2011)
Lanthanide Complexes of Polyacid Ligands derived from 2,6-bis(pyrazol-1-yl) pyridine, pyrazine, and 6,6'-bis(pyrazol-1-yl)-2, 2'-bipyridine: Synthesis and luminescence properties.
Rodriguez-Ubis JC, et al.
Helvetica Chimica Acta, 80(1), 86-96 (1997)
Yunpeng Ye et al.
Bioconjugate chemistry, 16(1), 51-61 (2005-01-20)
Synergistic multivalent interactions can amplify desired chemical or biological molecular recognitions. We report a new class of multicarboxylate-containing carbocyanine dye constructs for use as optical scaffolds that not only serve as fluorescent antennas but also participate in structural assembly of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service