Skip to Content
Merck
All Photos(3)

Key Documents

274135

Sigma-Aldrich

N,N-Dimethylacrylamide

99%, contains 500 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

2-Propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCON(CH3)2
CAS Number:
Molecular Weight:
99.13
Beilstein:
1742219
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

contains

500 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.473 (lit.)

bp

80-81 °C/20 mmHg (lit.)

density

0.962 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(=O)C=C

InChI

1S/C5H9NO/c1-4-5(7)6(2)3/h4H,1H2,2-3H3

InChI key

YLGYACDQVQQZSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacrylamide (DMAA) is a non-ionic monomer with high reactivity and low initiation temperature. It is commonly used as a monomer in the production of polymers and copolymers for various applications such as hydrogels, polymeric flocculants, biomedical and drug delivery applications, polymer coatings, smart coatings, adhesives, sensors, self-healing materials, and water treatment agents. The unique structure of DMMA can form a three-dimensional network with other hydrogels and enhance their water retention capacity. It is also used as a crosslinking agent in polymerization reactions.

Application

N, N-Dimethylacrylamide(DMAA) can be used in the following applications:
  • Synthesis of poly(N,N-dimethylacrylamide) (PDMAA) nanoparticles which are used in biomedical and drug delivery applications.
  • Use as a co-monomer in the synthesis of thermoresponsive polymers used in smart coatings, adhesives, and sensors.
  • As a reactive diluent in the manufacture of UV-cured coatings and inks.
  • Use as a co-monomer with polyethylene glycol dimethacrylate (PEGDMA) and a chitosan derivative to develop an injectable hydrogel that can be used as a bone tissue engineering matrix.
  • A series of DMMA-based hydrogels for the removal of heavy metals and organic dyes from polluted water. These hydrogels are highly efficient in removing cationic dyes due to electrostatic interaction and hydrogen bonding in the DMMA backbone.
  • Non-toxic luminescent carbon dot/poly(DMMA) nanocomposite via surface-initiated reversible addition-fragmentation chain transfer polymerization.
  • Polymer coating for quartz crystal microbalance(QCM) sensor. The poly(DMMA) gel acts as a sensing material to adsorb Au(III) ions from HCl aqueous solution and it is inactive to most other metal ions and organic compounds.

Features and Benefits

  • It is an appropriate precursor for copolymerization due to its low initiation temperature and high reactivity.
  • Its unique structure forms a 3D network and improves the water retention capacity of hydrogels.
  • It is highly soluble in water.
  • It can easily bind to colloidal particles to enhance the bridging effect which plays a key role in flocculation.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taka-Aki Asoh et al.
Scientific reports, 10(1), 17173-17173 (2020-10-16)
Hydrogel is an attractive material, but its application is limited due to its low mechanical strength. In this study, a tough composite gel could be prepared by synthesizing polymer particles within a polymer network having relatively loose cross-linking. Since the
Syed Ragib Safi et al.
Scientific reports, 9(1), 11909-11909 (2019-08-17)
Arsenic contamination of groundwater is a serious concern worldwide. The research gaps in removing arsenic are selectivity, regeneration and effective removal rate at neutral pH levels. In this study, we discussed the reasons of the high arsenic adsorption from groundwater
Graft copolymerization of N, N-Dimethylacrylamide to cellulose in homogeneous media using atom transfer radical polymerization for hemocompatibility
Yan L and Wei T
Journal of Biomedical Science and Engineering, 10(1), 37-37 (2008)
Yuriko Matsumura et al.
Polymers, 11(7) (2019-07-20)
In order to clarify the local environment during interpolymer complex formation between poly(carboxylic acids) and poly(acrylamide derivatives) with different N-substitutions, a fluorescence label technique was used. 3-(2-propenyl)-9-(4-N,N-dimethylaminophenyl) phenanthrene (VDP) was used as an intramolecular fluorescence probe. All polymers were synthesized
Mareike Wendorff et al.
Frontiers in immunology, 11, 1705-1705 (2020-09-10)
Human Leukocyte Antigen class II (HLA-II) molecules present peptides to T lymphocytes and play an important role in adaptive immune responses. Characterizing the binding specificity of single HLA-II molecules has profound impacts for understanding cellular immunity, identifying the cause of

Articles

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service