Skip to Content
Merck
All Photos(3)

Key Documents

495905

Sigma-Aldrich

Sodium hydrogencyanamide

98%

Synonym(s):

mono-Sodium cyanamide, Cyanamide monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCNHNa
CAS Number:
Molecular Weight:
64.02
Beilstein:
3624175
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

mp

>360 °C (lit.)

storage temp.

2-8°C

SMILES string

[Na]NC#N

InChI

1S/CHN2.Na/c2-1-3;/h2H;/q-1;+1

InChI key

MBEGFNBBAVRKLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesús García-Valdés et al.
Journal of inorganic biochemistry, 197, 110689-110689 (2019-05-18)
Bis-cations with two 2,3-diferrocenylcyclopropenium fragments 3a-d, and the cis-2-(1,2-diferrocenylvinyl)-2-imidazolinium tetrafluoroborates 4a, d or the cis-2-(1,2-diferrocenylvinyl)-3,4,5,6-tetrahydropyrimidin-2-ium tetrafluoroborates 4b, c were obtained by interactions of 2,3-diferrocenyl-1-ethoxycyclopropenium tetrafluoroborate 1 with bis-1,4-N,N-(2a, d) or bis-1,5-N,N-(2b, c) nucleophiles. The reactions of 3a-d with sodium azide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service