Skip to Content
Merck
All Photos(2)

Key Documents

438928

Sigma-Aldrich

Calcium cyanamide

technical grade

Synonym(s):

Calcium carbimide, Cyanamide calcium derivative, Cyanamide calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaNCN
CAS Number:
Molecular Weight:
80.10
Beilstein:
4124391
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

≥85% (perchloric acid titration)

form

solid

composition

Ca, 45%

mp

>300 °C (lit.)

SMILES string

[Ca]=NC#N

InChI

1S/CN2.Ca/c2-1-3;

InChI key

QFSRQFUIHVTIDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Determination of calcium cyanamide in air of workplace by spectrophotometry].
Wei Zhao et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 30(6), 474-476 (2012-08-31)
Ricardo Vergara et al.
Plant molecular biology, 79(1-2), 171-178 (2012-04-03)
It has been reported that dormancy-breaking compound hydrogen cyanamide (HC) stimulates the fermentative pathway and inhibits respiration in grapevine-buds, suggesting in this way, that a respiratory stress must be involved in the release of buds from dormancy. Here, we tested
Bobby D Nossaman et al.
Nitric oxide : biology and chemistry, 26(4), 223-228 (2012-04-03)
Responses to glyceryl trinitrate/nitroglycerin (GTN), S-nitrosoglutathione (GSNO), and sodium nitrite were compared in the intact chest rat. The iv injections of GTN, sodium nitrite, and GSNO produced dose-dependent decreases in pulmonary and systemic arterial pressures. In as much as cardiac
Carmen Harb et al.
Inorganic chemistry, 52(3), 1621-1630 (2013-01-24)
Nine [Ru(Tp)(dppe)L] complexes, where Tp is hydrotris(pyrazol-1-yl)borate, dppe is ethylenebis(diphenylphosphine), and L is (4-nitrophenyl)cyanamide (NO(2)pcyd(-)), (2-chlorophenyl)cyanamide (2-Clpcyd(-)), (3-chlorophenyl)cyanamide (3-Clpcyd(-)), (2,4-dichlorophenyl)cyanamide (2,4-Cl(2)pcyd(-)), (2,3-dichlorophenyl)cyanamide (2,3-Cl(2)pcyd(-)), (2,5-dichlorophenyl)cyanamide (2,5-Cl(2)pcyd(-)), (2,4,5-trichlorophenyl)cyanamide (2,4,5-Cl(3)pcyd(-)), (2,3,5,6-tetrachlorophenyl)cyanamide (2,3,5,6-Cl(4)pcyd(-)), and (pentachlorophenyl)cyanamide (Cl(5)pcyd(-)), and the dinuclear complex [{Ru(Tp)(dppe)}(2)(μ-adpc)], where adpc(2-) is
Mahmoud M El-Mas et al.
American journal of physiology. Heart and circulatory physiology, 302(3), H837-H844 (2011-12-14)
We have previously shown that ethanol microinjection into the rostral ventrolateral medulla (RVLM) elicits sympathoexcitation and hypertension in conscious spontaneously hypertensive rats (SHRs) but not in Wistar-Kyoto (WKY) rats. In this study, evidence was sought to implicate the oxidative breakdown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service