Skip to Content
Merck
All Photos(1)

Key Documents

493198

Sigma-Aldrich

Pentadecane-d32

98 atom % D, 98% (CP)

Synonym(s):

Deuterated pentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)13CD3
CAS Number:
Molecular Weight:
244.61
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

refractive index

n20/D 1.432 (lit.)

bp

270 °C (lit.)

mp

8-10 °C (lit.)

density

0.883 g/mL at 25 °C

mass shift

M+32

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2

InChI key

YCOZIPAWZNQLMR-UBTWNIDWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordan Lopez et al.
Journal of agricultural and food chemistry, 67(41), 11444-11453 (2019-10-09)
Innovative approaches to develop flavors with high sensory appeal are critical in encouraging increased consumer preference and adoption of low sodium foods. Gas chromatography-olfactometry, coupled with stable isotope dilution assays and sensory experiments, led to the identification of the odorants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service