Skip to Content
Merck
All Photos(1)

Key Documents

492949

Sigma-Aldrich

L-Leucine-2,3,3,4,5,5,5,5′,5′,5′-d10

98 atom % D

Synonym(s):

L-Leucine-C-d10, L-Leucine-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDCD2CD(NH2)CO2H
CAS Number:
Molecular Weight:
141.23
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]25/D +14.5°, c = 2 in 5 M HCl

mp

>300 °C (lit.)

mass shift

M+10

SMILES string

[2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[C@]([2H])(N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1D3,2D3,3D2,4D,5D

InChI key

ROHFNLRQFUQHCH-ZWFPVXGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Liu et al.
Molecular nutrition & food research, 59(11), 2107-2118 (2015-08-13)
The objective was to investigate the metabolome changes in female rats gavaged with partially purified cranberry procyanidins (PPCP) using (1) H NMR and UHPLC-Q-Orbitrap-HRMS metabolomics approaches, and to identify the contributing metabolites. Twenty-four female Sprague-Dawley rats were randomly separated into
Shaomin Zhao et al.
Molecular nutrition & food research, 64(11), e1901242-e1901242 (2020-04-14)
Methods to verify cranberry juice consumption are lacking. Predictive multivariate models built upon validated biomarkers may help to verify human consumption of a food using a nutrimetabolomics approach. A 21-day double-blinded, randomized, placebo-controlled, cross-over study was conducted among healthy young
Haiyan Liu et al.
Food & function, 11(3), 2466-2476 (2020-03-07)
The objectives of this research were to investigate urinary metabolome modifications and discover potential intake biomarkers in young women after cranberry juice consumption. Fifteen female college students were given either cranberry juice or apple juice for three days using a
Mohamad-Reza Aghanoori et al.
Molecular metabolism, 20, 149-165 (2018-12-14)
Diabetic sensorimotor polyneuropathy (DSPN) affects approximately half of diabetic patients leading to significant morbidity. There is impaired neurotrophic growth factor signaling, AMP-activated protein kinase (AMPK) activity and mitochondrial function in dorsal root ganglia (DRG) of animal models of type 1
Alberto Valdés et al.
Scientific reports, 11(1), 5058-5058 (2021-03-05)
Diabetic nephropathy (DN) is the leading cause of chronic kidney disease. Although hyperglycaemia has been determined as the most important risk factor, hypoxia also plays a relevant role in the development of this disease. In this work, a comprehensive metabolomic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service