Skip to Content
Merck
All Photos(1)

Key Documents

661562

Sigma-Aldrich

L-Methionine-(methyl-d3)

endotoxin tested, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

optical activity

[α]25/D +23.1°, c = 1 in 1 M HCl
[α]25/D +23.1°, c = 1 in 1 M HCl

mp

284 °C (dec.) (lit.)

suitability

endotoxin tested

mass shift

M+3

SMILES string

[2H]C([2H])([2H])SCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1D3

InChI key

FFEARJCKVFRZRR-OSIBIXDNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service