487686
4-Hydroxy-TEMPO-d17
97 atom % D, 95% (CP)
Synonym(s):
4-Hydroxy-2,2,6,6-tetramethylpiperidine-d17-1-oxyl, TEMPOL-d17
About This Item
Recommended Products
grade
for ESR-spectroscopy
Quality Level
isotopic purity
97 atom % D
Assay
95% (CP)
form
solid
mp
69-71 °C (lit.)
mass shift
M+17
storage temp.
2-8°C
SMILES string
[2H]C([2H])([2H])C1(N([O])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])(O)C1([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11H,5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D
InChI key
UZFMOKQJFYMBGY-DEMDKEALSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2 Oral
Target Organs
Liver,spleen
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service