92688
5,5-Dimethyl-1-pyrroline N-oxide
for ESR-spectroscopy
Synonym(s):
DMPO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for ESR-spectroscopy
Quality Level
Assay
≥98.0% (GC)
form
crystals
refractive index
n20/D 1.496 (lit.)
bp
75 °C/0.4 mmHg (lit.)
mp
25-29 °C (lit.)
density
1.015 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CC1(C)CCC=[N+]1[O-]
InChI
1S/C6H11NO/c1-6(2)4-3-5-7(6)8/h5H,3-4H2,1-2H3
InChI key
VCUVETGKTILCLC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
5,5-Dimethyl-1-pyrroline N-oxide is a reagent generally used either as a free-radical spin-trapping agent, or electrophilic component during the synthesis of pyrrolidine derivatives. It may also be considered as 1,3-dipole in cycloaddition processes.
Neuroprotective agent; nitric oxide spin trap. Used to study radicals formed by enzymatic acetaldehyde oxidation. Incubation of lymphocytes with DMPO decreased DNA damage by NiCl2.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
203.0 °F - closed cup
Flash Point(C)
95 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The journal of physical chemistry. A, 116(26), 7210-7218 (2012-06-07)
Radical forms of sulfur dioxide (SO(2)), sulfite (SO(3)(2-)), sulfate (SO(4)(2-)), and their conjugate acids are known to be generated in vivo through various chemical and biochemical pathways. Oxides of sulfur are environmentally pervasive compounds and are associated with a number
Free radical biology & medicine, 36(10), 1214-1223 (2004-04-28)
The detection of protein free radicals using the specific free radical reactivity of nitrone spin traps in conjunction with nitrone-antibody sensitivity and specificity greatly expands the utility of the spin trapping technique, which is no longer dependent on the quantum
Free radical biology & medicine, 53(4), 962-973 (2012-05-29)
Complex I is a critical site of O(2)(•-) production and the major host of reactive protein thiols in mitochondria. In response to oxidative stress, complex I protein thiols at the 51- and 75-kDa subunits are reversibly S-glutathionylated. The mechanism of
Journal of biochemistry, 150(2), 173-181 (2011-05-17)
The nicotinamide adenine dinucleotide (NADH)/nicotinamide adenine dinucleotide phosphate (NADPH) oxidase and the xanthine oxidase (XOD) systems generate reactive oxygen species (ROS). In the present study, to characterize the difference between the two systems, the kinetics of ROS generated by both
Nucleic acids research, 40(12), 5477-5486 (2012-03-06)
Oxidative stress-related damage to the DNA macromolecule produces lesions that are implicated in various diseases. To understand damage to DNA, it is important to study the free radical reactions causing the damage. Measurement of DNA damage has been a matter
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service