Skip to Content
Merck
All Photos(1)

Key Documents

480975

Sigma-Aldrich

Hydroxylamine-O-sulfonic acid

99.998%

Synonym(s):

(Aminooxy)(hydroxy)sulfane dioxide, (Aminooxy)sulfonic acid, Amidoperoxymonosulfuric acid, Amidosulfonic peracid, Aminomonopersulfuric acid, Ammonia-N-oxide-O-sulfonic acid betaine, Permonosulfamic acid, Sulfoperamidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NOSO3H
CAS Number:
Molecular Weight:
113.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.998%

mp

210 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

NOS(O)(=O)=O

InChI

1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4)

InChI key

DQPBABKTKYNPMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxylamine-O-sulfonic acid (HOSA), an O-substituted hydroxylamine, is a widely used aminating agent. It can behave as a nucleophile as well as an electrophile. The conversion of organoboranes obtained from hydroboration of olefins to amines using HOSA has been reported.

Application

Hydroxylamine-O-sulfonic acid (HOSA) may be used to synthesize (Z)-1H-purin-6(7H)-ylideneaminooxysulfonic acid.
HOSA may be used as an aminating reagent in the synthesis of:
  • 1-aminotetrazoles and 2-aminotetrazoles
  • N-aminopiperidine (NAPP)
  • 1H,1′ H-2,2′-biimidazole-1,1′-diamine

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A New Strategy for the Preparation of N-Aminopiperidine Using Hydroxylamine-O-Sulfonic Acid: Synthesis, Kinetic Modelling, Phase Equilibria, Extraction and Processes.
Labarthe E, et al.
Advances in Chemical Engineering and Science, 3(2) (2013)
Hydroxylamine-O-sulfonic Acid.
Erdik E and Saczewski J.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2013)
Amination of tetrazoles with hydroxylamine-O-sulfonic acid: 1-and 2-aminotetrazoles.
Raap R.
Canadian Journal of Chemistry, 47(19), 3677-3681 (1969)
Synthesis and molecular structure of (Z)-1H-purin-6-ylideneaminooxysulfonic acid: a possible secondary metabolite of adenine.
Saczewski J and Gdaniec M.
Heterocyclic Communications, 18(3), 109-112 (2012)
N-Trinitroethylamino functionalization of nitroimidazoles: a new strategy for high performance energetic materials.
Yin P, et al.
Journal of Material Chemistry A, 1(25), 7500-7510 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service