372455
Graphite, fluorinated, polymer
extent of labeling: >61 wt. % F
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
Quality Level
extent of labeling
>61 wt. % F
InChI
1S/C.F
InChI key
PRPAGESBURMWTI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Used in cathodic materials, lubricants, image recording, pressure-sensitive copying and carbon filament incandescent lamps.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pacing and clinical electrophysiology : PACE, 19(11 Pt 2), 1836-1840 (1996-11-01)
The reduction in pacemaker size coupled with the addition of more current demanding functions has motivated the development of batteries that can supply higher current densities at useful voltages than the lithium/iodine batteries in use today while retaining the volumetric
The journal of physical chemistry. A, 114(16), 5389-5396 (2010-04-08)
Carbon monofluoride (CF)(n) and graphane are two very different materials from the practical point of view, but the basic chemical motifs of these materials are closely related: both can be described as two-dimensional polycyclic (fluoro-/hydro-)carbons. However, the actual experimental data
Journal of magnetic resonance (San Diego, Calif. : 1997), 165(2), 230-236 (2003-12-04)
Rotational-echo double resonance (REDOR) is a magic-angle spinning technique for measuring heteronuclear dipolar couplings. Rotor-synchronized pi pulses recouple the dipolar interaction. The accuracy of a REDOR determination of distance or orientation depends totally on the quality of the dephased (recoupled)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service