Skip to Content
Merck
All Photos(2)

Key Documents

480142

Sigma-Aldrich

4-Phenoxyphenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4B(OH)2
CAS Number:
Molecular Weight:
214.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0%

mp

141-145 °C (lit.)

functional group

phenoxy

SMILES string

OB(O)c1ccc(Oc2ccccc2)cc1

InChI

1S/C12H11BO3/c14-13(15)10-6-8-12(9-7-10)16-11-4-2-1-3-5-11/h1-9,14-15H

InChI key

KFXUHRXGLWUOJT-UHFFFAOYSA-N

Application

4-Phenoxyphenylboronic acid can be used as a reactant:
  • In the Suzuki-Miyaura coupling reaction to synthesize aryl derivatives via C-C bond formation by reacting with different aryl halides over a palladium catalyst.[1][2]
  • To prepare 1-phenoxy-4-(trifluoromethyl)benzene via oxidative trifluoromethylation using Chan-Lam-type reaction conditions.[3]
  • To synthesize evobrutinib, a potent Bruton′s tyrosine kinase (BTK) inhibitor.[4]

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrazolo [3, 4-d] pyrimidines containing an extended 3-substituent as potent inhibitors of Lck-a selectivity insight
Burchat AF, et al.
Bioorganic & medicinal chemistry letters, 12(12), 1687-1690 (2002)
Room temperature aryl trifluoromethylation via copper-mediated oxidative cross-coupling
Senecal TD, et al.
The Journal of Organic Chemistry, 76(4), 1174-1176 (2011)
Efficacy and pharmacodynamic modeling of the BTK inhibitor evobrutinib in autoimmune disease models
Haselmayer P, et al.
Journal of Immunology, 202(10), 2888-2906 (2019)
Functionalization of pyrrolo [2, 3-d] pyrimidine by palladium-catalyzed cross-coupling reactions
Tumkevicius, S and Dodonova, J
Chemistry of Heterocyclic Compounds, 48(2), 258-279 (2012)
Imari Koike et al.
Planta, 251(3), 73-73 (2020-03-07)
Endogenous auxin determines the pattern of adventitious shoot formation. Auxin produced in the dominant shoot is transported to the internodal segment and suppresses growth of other shoots. Adventitious shoot formation is required for the propagation of economically important crops and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service