Skip to Content
Merck
All Photos(1)

Key Documents

477001

Sigma-Aldrich

2-Hexene (cis+trans)

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH3
CAS Number:
Molecular Weight:
84.16
Beilstein:
969179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

85%

refractive index

n20/D 1.395 (lit.)

bp

67-69 °C (lit.)

density

0.678 g/mL at 25 °C (lit.)

SMILES string

CCC\C=C\C

InChI

1S/C6H12/c1-3-5-6-4-2/h3,5H,4,6H2,1-2H3/b5-3+

InChI key

RYPKRALMXUUNKS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a mixture of cis- and trans-2-hexene. 1-hexene undergoes isomerization on 6080-mesh ZSM (Zeolite Socony Mobil)-5 zeolite to form cis- and trans-2-hexene as the predominant products.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The catalytic isomerization of 1-hexene on H-ZSM-5 zeolite: The effects of a shape-selective catalyst.
Abbot J, et al.
J. Catal., 92(2), 398-408 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service