Skip to Content
Merck
All Photos(1)

Key Documents

473197

Sigma-Aldrich

Poly(ethylene glycol) 4-nonylphenyl 3-sulfopropyl ether potassium salt

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

viscous liquid

refractive index

n20/D 1.473

bp

81 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

[K+].[S](=O)(=O)([O-])CCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOc1ccc(cc1)CCCCCCCCC

InChI

1S/C58H110O24S.K/c1-2-3-4-5-6-7-8-10-57-11-13-58(14-12-57)82-55-54-81-53-52-80-51-50-79-49-48-78-47-46-77-45-44-76-43-42-75-41-40-74-39-38-73-37-36-72-35-34-71-33-32-70-31-30-69-29-28-68-27-26-67-25-24-66-23-22-65-21-20-64-19-18-63-17-16-62-15-9-56-83(59,

InChI key

RCHNVNBUJMZZIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) 4-nonylphenyl 3-sulfopropyl ether potassium salt is an anionic PEG derivative that can be used as a soft polyoxyethylene sulfonate anion.

Application

Poly(ethylene glycol) 4-nonylphenyl 3-sulfopropyl ether potassium salt can be used as an anionic surfactant in the fabrication of hierarchical β-Ni(OH)2 microspheres and nickel oxide hollow spheres for supercapacitor applications.
Poly(ethylene glycol) 4-nonylphenyl 3-sulfopropyl ether potassium salt was also demonstrated to be effective for protecting thermostable DNA polymerase during amplification reactions conducted at a temperature ranging from about 40° C to greater than 100° C.
DNA polymerases, such as Taq, do become inactive over time at the high temperatures utilized in thermal cycling reactions such as polymerase chain reaction (PCR). It has been discovered that thermostable DNA polymerases could be protected from such heat inactivation by contacting the polymerase with poly(ethylene glycol) 4-nonylphenyl 3-sulfopropyl ether potassium salt during a thermal cycling process, and improve the specificity of amplification.

Physical form

~20 EO/mole

Legal Information

The composition of this product in combination with thermostable DNA polymerase (including, but not limited to, Taq polymerase) is covered by US Patent No. 7,972,828 and related foreign equivalents. In addition, use of this product may be covered by US Patent No. 8,404,464 and related foreign equivalents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalized ZnO nanoparticles with liquidlike behavior and their photoluminescence properties
Bourlinos AB, et al.
Small, 2(4), 513-516 (2006)
Reactive CeO2 nanofluids for UV protective films
Maniglia R, et al.
Journal of Colloid and Interface Science, 506(4), 346-354 (2017)
Rational synthesis of hierarchically porous NiO hollow spheres and their supercapacitor application
Yan X, et al.
Materials Letters, 95(14), 1-4 (2013)
Fabrication of novel hierarchical beta-Ni (OH) 2 and NiO microspheres via an easy hydrothermal process
Kuang D, et al.
The Journal of Physical Chemistry C, 113(14), 5508-5513 (2009)
Solvent-free protein liquids and liquid crystals.
Adam W Perriman et al.
Angewandte Chemie (International ed. in English), 48(34), 6242-6246 (2009-07-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service