Skip to Content
Merck
All Photos(2)

Key Documents

472360

Sigma-Aldrich

Poly(oxy-1,4-phenylenesulfonyl-1,4-phenylene)

melt index 6 g/10 min (380°C/2.16 kg)

Synonym(s):

Poly(1,4-phenylene ether-sulfone)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H8O3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

granular

melt index

6 g/10 min (380°C/2.16 kg)

ISO 1628

50 mL/g, in phenol/1,2-dichlorobenzene(lit.)

transition temp

Tg (DSC) 221 °C (onset)

solubility

aniline, dimethylacetamide and pyridine: soluble

density

1.37 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

InChI key

VPWNQTHUCYMVMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in electrical connectors, printed circuit boards, sterilizable equipment and appliance covers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Schneditz et al.
Clinical hemorheology and microcirculation, 58(3), 455-469 (2013-11-21)
It was the aim to examine the fluid flow in blood and dialysate compartments of highly permeable hollow fiber dialyzers where internal filtration contributes to solute removal but where excessive filtration bears a risk of cell activation and damage. Flow
Mario Bonomini et al.
Therapeutics and clinical risk management, 14, 1-9 (2018-01-04)
The success and the quality of hemodialysis therapy are mainly related to both clearance and biocompatibility properties of the artificial membrane packed in the hemodialyzer. Performance of a membrane is strongly influenced by its interaction with the plasma protein repertoire
Wenhong Zhu et al.
Journal of visualized experiments : JoVE, (66)(66), e4108-e4108 (2012-08-17)
Although human saliva proteome and peptidome have been revealed they were majorly identified from tryptic digests of saliva proteins. Identification of indigenous peptidome of human saliva without prior digestion with exogenous enzymes becomes imperative, since native peptides in human saliva
Shengqiang Nie et al.
Colloids and surfaces. B, Biointerfaces, 100, 116-125 (2012-07-06)
In this study, a novel triblock copolymer of poly (styrene-co-acrylic acid)-b-poly (vinyl pyrrolidone)-b-poly(styrene-co-acrylic acid) (P(St-co-AA)-b-PVP-b-P(St-co-AA)) is synthesized via reversible addition-fragmentation chain transfer (RAFT) polymerization, and used for the modification of blood contacting surface of polyethersulfone (PES) membrane to improve blood
Shogo Takamuku et al.
ChemSusChem, 6(2), 308-319 (2013-01-12)
The morphologies and proton-transport efficiencies of segmented copoly(arylene ether sulfone) ionomers that contain tetrasulfonated sequences are compared with the corresponding copolymers with disulfonated sequences. Tetrasulfonated 4,4'-bis[(4-chlorophenyl)sulfonyl]-1,1'-biphenyl (sBCPSBP) is synthesized by metalation and sulfination. This new monomer is then used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service