472034
Tripentylamine
98%, mixture of isomers
Synonym(s):
Triamylamine, mixed isomers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[CH3(CH2)4]3N
CAS Number:
Molecular Weight:
227.43
Beilstein:
1744528
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
1 mmHg ( 20 °C)
Quality Level
Assay
98%
refractive index
n20/D 1.436 (lit.)
bp
81-83 °C/0.2 mmHg (lit.)
density
0.782 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCCCCN(CCCCC)CCCCC
InChI
1S/C15H33N/c1-4-7-10-13-16(14-11-8-5-2)15-12-9-6-3/h4-15H2,1-3H3
InChI key
OOHAUGDGCWURIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The product is an isomeric mixture of tripentylamine (TPA). TPA is a tertiary amine. It forms charge transfer complexes with C60 in toluene. The stability constants of these complexes have been evaluated using the Benesi-Hildebrand equation.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Electron transfer with aliphatic substrates oxidation of cyclohexane with cobalt (III) ions alone and in the presence of oxygen.
Onopchenko A and Schulz JGD.
The Journal of Organic Chemistry, 38(21), 3729-3733 (1973)
Warwick et al.
Analytical chemistry, 72(16), 3960-3963 (2000-08-26)
The extraction of Tc-tripentylamine complexes onto C18 silica solid-phase extraction columns is evaluated for the selective removal of 99Tc from other isotopes. The Tc-amine complex is quantitatively extracted from sulfuric acid and can be recovered from the column by elution
A spectrophotometric study of the charge transfer complexation of [60] fullerene with some amines.
Mizyed S, et al.
Jordan Journal of Chemistry, 2(2), 145-153 (2007)
Dae Ho Lee et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 219-227 (2017-12-08)
Optimizing the colloidal state of polyamic acid (PAA) nanoparticles is essential for achieving a uniform and high-performance polyimide coating by electrophoretic deposition (EPD) on metal substrates of various shapes. In this paper, we report two important roles of the counterions
Michael K L Chu et al.
Diabetes technology & therapeutics, 17(4), 255-267 (2015-02-12)
An implantable, glucose-responsive insulin delivery microdevice was reported previously by our group, providing rapid insulin release in response to hyperglycemic events and efficacy in vivo over a 1-week period when implanted intraperitoneally in rats with diabetes. Herein, we focused on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service