Skip to Content
Merck
All Photos(2)

Key Documents

471623

Sigma-Aldrich

1,1,1-Trimethylhydrazinium iodide

97%

Synonym(s):

1,1,1-Trimethylhydrazine iodide, N,N,N-Trimethylhydrazinium iodide, Trimethylhydrazonium iodide (6CI)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NN(CH3)3I
CAS Number:
Molecular Weight:
202.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

225-230 °C (dec.) (lit.)

functional group

amine

SMILES string

[I-].C[N+](C)(C)N

InChI

1S/C3H11N2.HI/c1-5(2,3)4;/h4H2,1-3H3;1H/q+1;/p-1

InChI key

HLNPDVKKRBWQAA-UHFFFAOYSA-M

General description

1,1,1-Trimethylhydrazinium iodide ([(CH3)3N-NH2]I, TMHI) is widely employed as a vicarious nucleophilic substitution (VNS) reagent for various aromatic amination reactions. On reaction with silver salt, it affords the following salts: nitrate (([(CH3)3N-NH2][NO3]), perchlorate ([(CH3)3N-NH2][ClO4]), azide ([(CH3)3N-NH2][N3]), 5-amino-1H-tetrazolate ([(CH3)3N-NH2][H2N-CN4]) and sulfate ([(CH3)3N-NH2]2[SO4].2H2O).

Application

1,1,1-Trimethylhydrazinium iodide (TMHI) may be employed for the amination of 1-X-3,5-dinitrobenzenes.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir V Rozhkov et al.
The Journal of organic chemistry, 68(6), 2498-2501 (2003-03-15)
The amination of 1-X-3,5-dinitrobenzenes via the vicarious nucleophilic substitution of hydrogen (VNS) with 1,1,1-trimethylhydrazinium iodide (TMHI) in the presence of t-BuOK or NaOMe in DMSO was studied. It was observed (when X = OMe, OCH(2)CF(3), OCH(2)CF(2)CF(2)H, OPh) that the amination
Carles Miró Sabaté et al.
Chemistry, an Asian journal, 7(9), 2080-2089 (2012-06-29)
1,1,1-Trimethylhydrazinium iodide ([(CH(3))(3)N-NH(2)]I, 1) was reacted with a silver salt to form the corresponding nitrate ([(CH(3))(3)N-NH(2)][NO(3)], 2), perchlorate ([(CH(3))(3)N-NH(2)][ClO(4)], 3), azide ([(CH(3))(3)N-NH(2)][N(3)], 4), 5-amino-1H-tetrazolate ([(CH(3))(3)N-NH(2)][H(2)N-CN(4)], 5), and sulfate ([(CH(3))(3)N-NH(2)](2)[SO(4)]·2H(2)O, 6·2H(2)O) salts. The metathesis reaction of compound 6·2H(2)O with barium salts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service