Skip to Content
Merck
All Photos(1)

Key Documents

471054

Sigma-Aldrich

Lignosulfonic acid calcium salt

average Mw ~18,000, average Mn ~2,500

Synonym(s):

Calcium ligninsulfonate, Lignin calcium sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn ~2,500
average Mw ~18,000

Quality Level

composition

Ca, 5 wt. %

impurities

16 wt. % reducing sugars
5 wt. %

solubility

H2O: soluble

SMILES string

[Ca+2].[S](=O)(=O)([O-])CCCc1cc(c(cc1)OC(C[S](=O)(=O)O)Cc2c(c(ccc2)OC)[O-])OC

InChI

1S/C20H26O10S2.Ca/c1-28-18-7-3-6-15(20(18)21)12-16(13-32(25,26)27)30-17-9-8-14(11-19(17)29-2)5-4-10-31(22,23)24;/h3,6-9,11,16,21H,4-5,10,12-13H2,1-2H3,(H,22,23,24)(H,25,26,27);/q;+2/p-2

InChI key

RYAGRZNBULDMBW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Granulating agent for limestone and ores, compaction aid for coal and metal wastes, and dispersing agent for gypsum and clay slurries.

Features and Benefits

Compatible with anionic and nonionic dispersants and wetting agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reid B Grigg et al.
Journal of colloid and interface science, 279(1), 36-45 (2004-09-24)
This paper describes adsorption and desorption studies carried out with calcium lignosulfonate (CLS) on Berea sandstone. Circulation experiments were performed to determine CLS adsorption isotherms and the effects of CLS concentration, temperature, salinity, brine hardness, and injection rate on adsorption
Shu-Juan Zhang et al.
Chemosphere, 57(9), 1181-1187 (2004-10-27)
Gamma-ray irradiation was proven to be a promising means for the removal of calcium lignosulfonate (CaLS). At a dose rate of 55Gy min(-1), over 90% of CaLS was mineralized to CO(2), H(2)O and sulfates within 3-d irradiation. The degradation of
Weixing Wang et al.
Chemical communications (Cambridge, England), 48(95), 11638-11640 (2012-10-18)
Lignosulfonates, which are byproducts of the pulp and paper industry, can be used as promoters for the formation of methane hydrates with a high capacity up to 170 v/v and a high formation rate.
Sangobtip Pongstabodee et al.
Waste management (New York, N.Y.), 28(3), 475-483 (2007-05-12)
The aim of this research was to separate the different plastics of a mixed post-consumer plastic waste by the combination of a three-stage sink-float method and selective flotation. By using the three-stage sink-float method, six mixed-plastic wastes, belonging to the
H R Mansfield et al.
Journal of dairy science, 77(4), 1070-1083 (1994-04-01)
Four Holstein cows were used in a 4 x 4 Latin square design to investigate the effects of soybean hulls and lignosulfonate-treated soybean meal on ruminal fermentation and nutrient passage to the duodenum. Diets contained 32% corn silage, 19.8% alfalfa-grass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service