Skip to Content
Merck
All Photos(1)

Key Documents

466964

Sigma-Aldrich

4′-Fluoro-2′-hydroxyacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OH)COCH3
CAS Number:
Molecular Weight:
154.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

31-35 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC(=O)c1ccc(F)cc1O

InChI

1S/C8H7FO2/c1-5(10)7-3-2-6(9)4-8(7)11/h2-4,11H,1H3

InChI key

HLTBTUXAMVOKIH-UHFFFAOYSA-N

General description

4′-Fluoro-2′-hydroxyacetophenone is a substituted acetophenone derivative. Biological Baeyer-Villiger oxidation of 4′-fluoro-2′-hydroxyacetophenone to 4-fluorocatechol by using whole cells of Pseudomonas fluorescens ACB has been reported. Its crystals belong to the monoclinic crystal system and space group P21/n.

Application

4′-Fluoro-2′-hydroxyacetophenone may be used in the preparation of series of 4′-fluoro-2′-hydroxychalcones, via aldol condensation with substituted aldehydes followed by cyclization with hydrazine hydrate.
Used recently in the preparation of medicinally active benzo[b]furans and thiophenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Moonen et al.
Journal of industrial microbiology & biotechnology, 26(1-2), 35-42 (2001-09-11)
The biological Baeyer-Villiger oxidation of acetophenones was studied by 19F nuclear magnetic resonance (NMR). The 19F NMR method was used to characterise the time-dependent conversion of various fluorinated acetophenones in either whole cells of Pseudomonas fluorescens ACB or in incubations
Khaled R A Abdellatif et al.
Journal of enzyme inhibition and medicinal chemistry, 30(3), 484-491 (2014-09-10)
In an effort to develop safe and potent anti-inflammatory agents, a series of novel 4'-fluoro-2'-hydroxychalcones 5a-d and their dihydropyrazole derivatives 6a-d was prepared. It was synthesized via aldol condensation of 4'-fluoro-2'-hydroxyacetophenone with appropriately substituted aldehydes followed by cyclization with hydrazine
4'-Fluoro-2'-hydroxyacetophenone.
Rizal MR and Ng SW.
Acta Crystallographica Section E, Structure Reports Online, 64(5), o916-o916 (2008)
Journal of Heterocyclic Chemistry, 30, 445-445 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service