Skip to Content
Merck
All Photos(1)

Key Documents

74880

Sigma-Aldrich

4-Octanol

≥97.0% (GC)

Synonym(s):

Butyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH2CH2CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719296
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.426

bp

174-176 °C (lit.)

density

0.818 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCC(O)CCC

InChI

1S/C8H18O/c1-3-5-7-8(9)6-4-2/h8-9H,3-7H2,1-2H3

InChI key

WOFPPJOZXUTRAU-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Winters et al.
Journal of applied microbiology, 127(3), 778-793 (2019-06-19)
Yeast and bacterial communities inhabit a sourdough starter to make artisanal bread. This study shows whether the interactions of micro-organisms derived from Australian sourdough starters provide some of the positive flavour, and aroma properties to bread by using defined sourdough
Qi Wang et al.
Food research international (Ottawa, Ont.), 140, 109995-109995 (2021-03-03)
As the main raw material for commercial Wuliangye-flavor liquor, Wuliangye-flavor raw liquor (WFRL) plays an important role in the formation of flavor components. Aroma components and microbial community diversity of 4 WFRLs produced by the same fermenting cellar were investigated
Ling You et al.
World journal of microbiology & biotechnology, 37(2), 26-26 (2021-01-12)
The role of the yeast community in Chinese strong-flavor baijiu fermentation was investigated by culture-independent and culture-dependent methods based on 26S rDNA sequence analysis. 92 yeast species belonging to 54 genera were found by Illumina sequencing during the fermentation of
Yun Xiong et al.
Journal of the science of food and agriculture, 100(3), 978-985 (2019-10-17)
Sorghum grain is rich in phenolic compounds and has the potential to be developed into functional beverages such as sorghum grain tea, in which the health benefits and flavour are the important quality attributes to be considered in tea product
Hehe Li et al.
Food chemistry, 284, 100-107 (2019-02-13)
Key odorants of Guojing sesame-flavor Baijiu were identified using the concept of molecular sensory science, including aroma extract dilution analysis (AEDA) based on gas chromatography-olfactometry (GC-O) combined with gas chromatography-mass spectrometry (GC-MS). A total of 509 volatile compounds were detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service