Skip to Content
Merck
All Photos(1)

Key Documents

359963

Sigma-Aldrich

Boron trifluoride dihydrate

96%

Synonym(s):

Boron fluoride dihydrate, Trifluoroborane dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · 2H2O
CAS Number:
Molecular Weight:
103.84
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

refractive index

n20/D 1.316 (lit.)

density

1.636 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].FB(F)F

InChI

1S/BF3.2H2O/c2-1(3)4;;/h;2*1H2

InChI key

MJCYPBSRKLJZTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Boron trifluoride dihydrate can be used as:
  • An activator in Pechmann condensation to synthesize 4-methylcoumarin derivatives by the reaction between substituted phenols and methyl acetoacetate.
  • An initiator for the polymerization of benzoxazine.
  • A catalyst in the regioselective Fries rearrangement of phenolic esters to give hydroxyphenyl alkyl/aryl ketones.
  • A catalyst to prepare 8-hydroxymethyltricyclene acetate by the reaction between camphene and formaldehyde in the presence of methylene chloride-acetic anhydride solvent.

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terpene-formaldehyde reactions. II. d-Limonene
Blomquist AT and Himics RJ
The Journal of Organic Chemistry, 33(3), 1156-1159 (1968)
Cationic polymerization of benzoxazine monomers by boron trifluoride complex.
Cid JA, et al.
Polymers & Polymer Composites, 7(6), 409-420 (1999)
BF3-H2O catalyzed Fries rearrangement of phenolic esters
Prakash GKS, et al.
Catalysis Letters, 114(1), 24-29 (2007)
Pechmann reaction promoted by boron trifluoride dihydrate.
Stoyanov E and Mezger J.
Molecules (Basel), 10(7), 762-766 (2005)
Diego Méndez et al.
Biochemical pharmacology, 183, 114341-114341 (2020-11-17)
Platelets are the smallest blood cells, and their activation (platelet cohesion or aggregation) at sites of vascular injury is essential for thrombus formation. Since the use of antiplatelet therapy is an unsolved problem, there are now focused and innovative efforts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service