Skip to Content
Merck
All Photos(3)

Key Documents

456055

Sigma-Aldrich

Camphene

95%

Synonym(s):

(±)-Camphene, 2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, 2,2-Dimethyl-3-methylenenorbornane, 2,2-Dimethyl-3-methylidenebicyclo[2.2.1]heptane, 2-Methylene-3,3-dimethylbicyclo[2.2.1]heptane, 3,3-Dimethyl-2-methylenenorbornane, 3,3-Dimethyl-2-methylenenorcamphane, DL-Camphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

bp

159-160 °C (lit.)

mp

48-52 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

SMILES string

[H][C@]12CC[C@]([H])(C1)C(C)(C)C2=C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m0/s1

InChI key

CRPUJAZIXJMDBK-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Camphene is abicyclic monoterpene and a constituent of many essential oils derived fromvarious plants such as Alpinia speciosa, Xanthium strumariumleaves, needles of Pinus uncinata, and Pinus uliginosa.(5)

Application

Camphene can be used as a reactant to synthesize:
  • Isobornyl carboxylates by silica-supported tungstophosphoric acid-catalyzed liquid-phase esterification of C2-C6 fatty acids.
  • Hydroaminated camphene via intermolecular anti-Markovnikov hydroamination reaction with N-hydroxyphthalimide and triethyl phosphite in the presence of dilauroyl peroxide as an initiator.
  • Camphene oxide via methyltrioxorhenium-catalyzed epoxidation in the presence of H2O2 as an oxidant and pyrazole as a Lewis base adduct.
  • Isobornyl alkyl ethers using alcohols via cation exchange resin-catalyzed alkoxylation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dendritic growth kinetics and structure II. Camphene.
Rubinstein ER and Glicksman ME.
Journal of Crystal Growth, 112(1), 97-110 (1991)
Radosław Bonikowski et al.
Natural product communications, 10(2), 371-373 (2015-04-30)
The compositions of mountain pine (Pinus uncinata) and peat-bog pine (P. uliginosa) needle essential oils were investigated. Enantiomeric compositions of selected monoterpene hydrocarbons were also examined. Respectively, fifty-three and seventy-six components of the essential oils were identified using GC-MS and
S-J Hong et al.
Acta biomaterialia, 5(5), 1725-1731 (2009-01-15)
Microparticulates are useful for directly filling defective tissues as well as for delivering cells and bioactive molecules in regenerative medicine. This paper reports on the production of bioactive ceramic microspheres with an interconnected macropore structure. The sol-gel derived calcium silicate
Seok-Jung Hong et al.
Macromolecular bioscience, 9(7), 639-645 (2009-02-20)
PCL microspheres featuring a macroporous morphology and a bone-bioactive surface have been prepared. 'Camphene' was introduced to generate pores within the microsphere network. The pore size was variable from a few to tens to hundreds of microm depending on the
Young J Hong et al.
Organic & biomolecular chemistry, 8(20), 4589-4600 (2010-08-21)
Based on quantum chemical studies, mechanisms to form bornyl diphosphate from geranyl diphosphate are suggested. While bornyl diphosphate is usually proposed to be generated via combination of the pyrophosphate group with a secondary bornyl cation, quantum chemical computations indicate that

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service