Skip to Content
Merck
All Photos(1)

Key Documents

13112

Sigma-Aldrich

Magnesium

powder, ≥99%

Synonym(s):

Magnesium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12141502
eCl@ss:
38110201
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

Quality Level

Assay

≥99%

form

powder

autoignition temp.

950 °F

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

impurities

≤0.05% insoluble in HCl

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

cation traces

Fe: ≤500 mg/kg

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium (powder) can be used as a reducing agent to synthesize:
  • Titanium alloy hydride powder by the reduction of upgraded titania slag (UGS) under hydrogen atmosphere.      
  • Ultrafine metallic iron (bcc-Fe) powders from hematite (α-Fe2O3) by ball-milling process.

It can also be used as a reducing agent in ruthenium(III) complex catalyzed atom-transfer radical addition (ATRA) and cyclization (ATRC) reactions. Magnesium does not generate free radicals but it generates and regenerates the catalytically active ruthenium(II) species.

Analysis Note

appearance : silver-gray

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched polyalkenylsilanes by hydrosilylation with platinum catalysts. I. Polymerization
Drohmann C, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 38(4), 741-751 (2000)
Katrin Thommes et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(24), 6899-6907 (2007-06-05)
A new catalytic procedure for atom-transfer radical addition (ATRA) and cyclization (ATRC) reactions is described. The combination of the ruthenium(III) complex [RuCl(2)Cp*(PPh3)] (Cp*: pentamethylcyclopentadienyl) with magnesium allows these reactions to be performed under mild conditions with high efficiency. In most
Ultrafine spinel powders by flame spray pyrolysis of a magnesium aluminum double alkoxide.
Bickmore CR, et al.
Journal of the American Chemical Society, 79(5), 1419-1423 (1996)
Uttley C.
Magnesium, 4-4 null
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service