Skip to Content
Merck

455903

Sigma-Aldrich

Potassium tetrafluoroborate

≥99.99% trace metals basis

Synonym(s):

Potassium borofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBF4
CAS Number:
Molecular Weight:
125.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay:
≥99.99% trace metals basis
form:
solid
Pricing and availability is not currently available.

Quality Level

Assay

≥99.99% trace metals basis

form

solid

impurities

<100.0 ppm Trace Metal Analysis

mp

530 °C (lit.)

density

2.505 g/mL at 25 °C (lit.)

SMILES string

[K+].F[B-](F)(F)F

InChI

1S/BF4.K/c2-1(3,4)5;/q-1;+1

InChI key

AKEBROIVCDHVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
379824451622278955
assay

≥99.99% trace metals basis

assay

99.99% trace metals basis

assay

99.99% trace metals basis

assay

96%

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

mp

530 °C (lit.)

mp

-

mp

293-300 °C (dec.) (lit.)

mp

530 °C (lit.)

density

2.505 g/mL at 25 °C (lit.)

density

-

density

-

density

2.505 g/mL at 25 °C (lit.)

impurities

<100.0 ppm Trace Metal Analysis

impurities

≤150.0 ppm Trace Metal Analysis

impurities

≤150.0 ppm Trace Metal Analysis

impurities

-

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia Kaluzhny et al.
Alternatives to laboratory animals : ATLA, 43(2), 101-127 (2015-05-23)
The 7th Amendment to the EU Cosmetics Directive and the EU REACH Regulation have reinforced the need for in vitro ocular test methods. Validated in vitro ocular toxicity tests that can predict the human response to chemicals, cosmetics and other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service