451762
Adipic acid-d10
98 atom % D
Synonym(s):
Deuterated adipic acid, Hexanedioic acid-d10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
DO2C(CD2)4CO2D
CAS Number:
Molecular Weight:
156.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % D
Quality Level
form
solid
bp
265 °C/100 mmHg (lit.)
mp
151-154 °C (lit.)
mass shift
M+10
SMILES string
[2H]OC(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O[2H]
InChI
1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1D2,2D2,3D2,4D2/hD2
InChI key
WNLRTRBMVRJNCN-YQMDOBQVSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
384.8 °F - closed cup
Flash Point(C)
196 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service