Skip to Content
Merck
All Photos(1)

Key Documents

45150

Sigma-Aldrich

Pamoic acid

≥97.0% (T)

Synonym(s):

1,1′-Methylene-bis(2-hydroxy-3-naphthoic acid), 4,4′-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16O6
CAS Number:
Molecular Weight:
388.37
Beilstein:
901319
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

mp

≥300 °C (dec.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cc2ccccc2c(Cc3c(O)c(cc4ccccc34)C(O)=O)c1O

InChI

1S/C23H16O6/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29/h1-10,24-25H,11H2,(H,26,27)(H,28,29)

InChI key

WLJNZVDCPSBLRP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Multi-component hydrogen-bonding assembly of a pharmaceutical agent pamoic acid with piperazine or 4, 4'-bipyridyl: A channel hydrated salt with multiple-helical motifs vs a bimolecular cocrystal.
Du M, et al.
Crystal Growth & Design, 9(4), 1655-1657 (2009)
Pamoic acid determined from powder diffraction data.
Haynes DA, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(3), o1170-o1172 (2006)
A series of four-connected entangled metal-organic frameworks assembled from pamoic acid and pyridine-containing ligands: interpenetrating, self-penetrating, and supramolecular isomerism.
MLA Wang S, et al.
Crystal Growth & Design, 12(1), 79-92 (2011)
Chad Deisenroth et al.
Molecular and cellular biology, 30(16), 3981-3993 (2010-06-16)
The ever-expanding knowledge of the role of p53 in cellular metabolism, apoptosis, and cell cycle control has led to increasing interest in defining the stress response pathways that regulate Mdm2. In an effort to identify novel Mdm2 binding partners, we
R W Roos et al.
Journal of chromatography, 370(3), 403-418 (1986-12-24)
Triethylamine (TEA) was evaluated as a competing base for the retention control and peak shape improvement in the reversed-phase high-performance liquid chromatographic (RP-HPLC) analysis of selected acidic, basic, and neutral drugs. The effects of this amine on the capacity factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service