Skip to Content
Merck
All Photos(2)

Key Documents

450359

Sigma-Aldrich

N-Boc-aniline

97%

Synonym(s):

tert-Butyl-N-phenylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCO2C(CH3)3
CAS Number:
Molecular Weight:
193.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

133-137 °C (lit.)

SMILES string

CC(C)(C)OC(=O)Nc1ccccc1

InChI

1S/C11H15NO2/c1-11(2,3)14-10(13)12-9-7-5-4-6-8-9/h4-8H,1-3H3,(H,12,13)

InChI key

KZZHPWMVEVZEFG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

SNAr reactions of benzaldimines: a concise synthesis of substituted phenanthridines.
Reuter DC, et al.
Tetrahedron Letters, 35(28), 4899-4902 (1994)
Directed ortho lithiation of phenylcarbamic acid 1, 1-dimethylethyl ester (N-BOC-aniline). Revision and improvements.
Stanetty P, et al.
The Journal of Organic Chemistry, 57(25), 6833-6837 (1992)
Novel practical deprotection of N-Boc compounds using fluorinated alcohols.
Choy J, et al.
Synthetic Communications, 38(21), 3840-3853 (2008)
Catalyst-free water-mediated N-Boc deprotection.
Wang G, et al.
Tetrahedron Letters, 50(13), 1438-1440 (2009)
Total synthesis of the polycyclic fungal metabolite (+/-)-communesin F.
Peng Liu et al.
Angewandte Chemie (International ed. in English), 49(11), 2000-2003 (2010-02-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service